siRNA Details
Gene Symbol: GOLGA2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027001.1 | 3 | 582 | |||
GenBank mRNA | AK307326.1 | - | 4 | 639 | ||
AK309056.1 | - | 4 | 506 | |||
AK311265.1 | - | 3 | 504 | |||
BC054509.1 | - | 3 | 399 | |||
BC071951.1 | - | 3 | 539 | |||
BC096759.1 | - | 3 | 539 |
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026811.3 | 4 | 688 | |||
NR_033936.3 | 3 | 599 | ||||
NR_103496.2 | 3 | 549 | ||||
GenBank mRNA | AK309732.1 | - | 3 | 550 | ||
AK310769.1 | - | 3 | 502 | |||
AK311170.1 | - | 3 | 509 | |||
BC007811.2 | - | 4 | 612 | |||
BC065260.1 | - | 4 | 605 | |||
BC072390.1 | - | 3 | 552 |
Gene Symbol: GOLGA2P2Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001555.2 | 8 | 961 | |||
GenBank mRNA | AF332229.1 | - | 8 | 960 |
Gene Symbol: GOLGA2P3Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002195.2 | 8 | 961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
