siRNA Details
Gene Symbol: GOLGA2P2Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001555.2 | 8 | 937 | |||
GenBank mRNA | AF332229.1 | - | 8 | 936 |
Gene Symbol: GOLGA2P3Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002195.2 | 8 | 937 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
