siRNA Details
Gene Symbol: DTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286229.1 | NP_001273158.1 | 13 | 2632 | ||
NM_001286230.1 | NP_001273159.1 | 14 | 2736 | |||
NM_016448.3 | NP_057532.3 | 15 | 2862 | |||
XM_011509614.1 | XP_011507916.1 | 15 | 2566 | |||
GenBank mRNA | AF195765.1 | - | 15 | 2626 | ||
AF345896.1 | - | 15 | 2647 | |||
AK000742.1 | - | 15 | 2680 | |||
AK001261.1 | - | 6 | 1669 | |||
AK027651.1 | - | 13 | 2339 | |||
BC033297.1 | - | 15 | 2603 | 4342989 | ||
BC033540.1 | - | 15 | 2644 | 4826434 |
