siRNA Details
Gene Symbol: MAP1LC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022818.4 | NP_073729.1 | 4 | 455 | ||
GenBank mRNA | AF087871.1 | - | 4 | 314 | ||
AF183417.1 | - | 4 | 314 | |||
AF303888.1 | - | 4 | 260 | |||
AK025556.1 | - | 4 | 306 | |||
AK290956.1 | - | 4 | 337 | |||
AK310224.1 | - | 5 | 422 | |||
AL834298.1 | - | 4 | 224 | |||
BC018634.2 | - | 4 | 293 | 3623259 | ||
BC041874.1 | - | 4 | 850 | 5271912 | ||
BC045759.1 | - | 5 | 796 | 4828857 | ||
BC048009.1 | - | 4 | 339 | |||
BC067797.1 | - | 4 | 306 | 5276841 | ||
BG771621.1 | - | 4 | 440 | |||
DA577457.1 | - | 4 | 335 | |||
DB055844.1 | - | 4 | 455 | |||
JN663879.1 | - | 4 | 246 |
Gene Symbol: MAP1LC3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085481.1 | NP_001078950.1 | 2 | 376 | ||
XM_011538652.2 | XP_011536954.1 | 2 | 549 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
