siRNA Details
Gene Symbol: KIAA1143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320334.1 | NP_001307263.1 | 2 | 231 | ||
NM_020696.3 | NP_065747.1 | 2 | 231 | |||
GenBank mRNA | AB032969.1 | - | 1 | 38 | ||
AK289549.1 | - | 2 | 203 | |||
BC008468.1 | - | 2 | 206 | 4277104 | ||
BC016790.1 | - | 2 | 165 | 4102473 | ||
BC070360.1 | - | 2 | 161 | |||
CD049061.1 | - | 2 | 228 | |||
DA183520.1 | - | 2 | 224 | |||
DA192476.1 | - | 2 | 206 | |||
DB238094.1 | - | 2 | 187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
