siRNA Details
Gene Symbol: KIAA1143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320334.1 | NP_001307263.1 | 3 | 506 | ||
NM_020696.3 | NP_065747.1 | 3 | 451 | |||
GenBank mRNA | AB032969.1 | - | 2 | 258 | ||
AK289549.1 | - | 3 | 423 | |||
BC008468.1 | - | 3 | 426 | 4277104 | ||
BC016790.1 | - | 3 | 385 | 4102473 | ||
BC070360.1 | - | 3 | 381 | |||
CD049061.1 | - | 3 | 448 | |||
DA183520.1 | - | 3 | 444 | |||
DA192476.1 | - | 3 | 426 | |||
DB238094.1 | - | 3 | 462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
