siRNA Details
Gene Symbol: KIAA1143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320334.1 | NP_001307263.1 | 3 | 529 | ||
NM_020696.3 | NP_065747.1 | 3 | 474 | |||
GenBank mRNA | AB032969.1 | - | 2 | 281 | ||
AK289549.1 | - | 3 | 446 | |||
BC008468.1 | - | 3 | 449 | 4277104 | ||
BC016790.1 | - | 3 | 408 | 4102473 | ||
BC070360.1 | - | 3 | 404 | |||
CD049061.1 | - | 3 | 471 | |||
DA183520.1 | - | 3 | 467 | |||
DA192476.1 | - | 3 | 449 | |||
DB238094.1 | - | 3 | 485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
