siRNA Details
Gene Symbol: UNC93B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030930.3 | NP_112192.2 | 11 | 1692 | ||
XM_011545290.1 | XP_011543592.1 | 9 | 1302 | |||
XM_011545291.2 | XP_011543593.1 | 10 | 1584 | |||
GenBank mRNA | AJ271326.1 | - | 11 | 1651 | ||
AY007125.1 | - | 10 | 1479 | |||
BC092472.1 | - | 11 | 1660 | 30348121 | ||
BC101568.1 | - | 11 | 1636 | 8069074 | ||
BC105104.1 | - | 11 | 1636 | 8144107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
