siRNA Details
Gene Symbol: STAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282716.1 | NP_001269645.1 | 10 | 1211 | ||
NM_001282717.1 | NP_001269646.1 | 10 | 1292 | |||
NM_001282718.1 | NP_001269647.1 | 8 | 1085 | |||
NM_012447.3 | NP_036579.2 | 10 | 1292 | |||
XM_011515742.1 | XP_011514044.1 | 10 | 1259 | |||
XM_017011683.1 | XP_016867172.1 | 10 | 1139 | |||
XM_017011684.1 | XP_016867173.1 | 10 | 1259 | |||
XM_017011685.1 | XP_016867174.1 | 10 | 1372 | |||
XM_017011686.1 | XP_016867175.1 | 10 | 1139 | |||
XM_017011687.1 | XP_016867176.1 | 8 | 965 | |||
GenBank mRNA | AJ007798.2 | - | 10 | 1212 | ||
AK292394.1 | - | 10 | 1119 | |||
AK302587.1 | - | 7 | 703 | |||
AK302693.1 | - | 8 | 908 | |||
AL833816.1 | - | 9 | 1383 | |||
BC140932.1 | - | 10 | 1051 | 9056744 | ||
BC146806.1 | - | 10 | 1117 | 8860345 |
Gene Symbol: STAG3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040583.2 | 5 | 576 | |||
GenBank mRNA | AK289466.1 | - | 6 | 545 | ||
AK291820.1 | - | 3 | 216 |
Gene Symbol: STAG3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309007.1 | - | 5 | 407 | ||
BI753182.1 | - | 5 | 397 |
Gene Symbol: STAG3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040582.2 | 4 | 595 | |||
GenBank mRNA | AK303592.1 | - | 4 | 256 | ||
AK316130.1 | - | 4 | 458 | |||
AY313779.1 | - | 4 | 450 | |||
BC066937.1 | - | 4 | 328 | 4826886 | ||
BG472630.1 | - | 4 | 422 | |||
BI829811.1 | - | 4 | 330 | |||
DA505391.1 | - | 4 | 467 | |||
DA555040.1 | - | 4 | 477 | |||
DC395490.1 | - | 4 | 378 | |||
DC410426.1 | - | 4 | 256 |
Gene Symbol: TRIM74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012197.1 | XP_016867686.1 | 4 | 467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
