siRNA Details
Gene Symbol: STAG3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040583.2 | 6, 7 | 782 | |||
GenBank mRNA | AK289466.1 | - | 7, 8 | 751 | ||
AK291820.1 | - | 4, 5 | 422 | |||
BC119762.1 | - | 2, 3 | 227 | |||
BG829469.1 | - | 4, 5 | 544 |
Gene Symbol: STAG3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040584.2 | 4, 5 | 526 | |||
GenBank mRNA | AK303473.1 | - | 5, 6 | 488 | ||
AK307228.1 | - | 5, 6 | 605 | |||
AK309007.1 | - | 6, 7 | 613 | |||
BC002581.2 | - | 5, 6 | 476 | |||
BC119761.1 | - | 2, 3 | 227 | 40119328 | ||
BG537494.1 | - | 4, 5 | 533 | |||
BI753182.1 | - | 6, 7 | 603 | |||
CB956482.1 | - | 4, 5 | 505 |
Gene Symbol: STAG3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040582.2 | 5, 6 | 801 | |||
GenBank mRNA | AK303592.1 | - | 5, 6 | 462 | ||
AY313779.1 | - | 5, 6 | 656 | |||
BC030509.1 | - | 2, 3 | 186 | |||
BC066937.1 | - | 5, 6 | 534 | 4826886 | ||
BC108680.1 | - | 4, 5 | 544 | |||
BG472630.1 | - | 5, 6 | 628 | |||
BI829811.1 | - | 5, 6 | 536 | |||
BM007884.1 | - | 4, 5 | 471 | |||
DC410426.1 | - | 5, 6 | 462 |
Gene Symbol: TRIM74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012197.1 | XP_016867686.1 | 5, 6 | 673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
