siRNA Details
Gene Symbol: STAG3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040583.2 | 8, 9 | 957 | |||
GenBank mRNA | AK289466.1 | - | 9, 10 | 926 | ||
BC119762.1 | - | 4, 5 | 402 |
Gene Symbol: STAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ007798.2 | - | 13, 14 | 1593 |
Gene Symbol: STAG3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040584.2 | 6, 7 | 701 | |||
GenBank mRNA | BC002581.2 | - | 7, 8 | 651 | ||
BC119761.1 | - | 4, 5 | 402 | 40119328 | ||
CB956482.1 | - | 6, 7 | 680 |
Gene Symbol: STAG3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040582.2 | 7, 8 | 976 | |||
GenBank mRNA | AL137492.1 | - | 2, 3 | 147 | ||
BC066937.1 | - | 7, 8 | 709 | 4826886 | ||
BC108680.1 | - | 6, 7 | 719 | |||
BI829811.1 | - | 7, 8 | 711 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
