siRNA Details
Gene Symbol: H2AFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012412.4 | NP_036544.1 | 4 | 367 | ||
NM_138635.3 | NP_619541.1 | 4 | 367 | |||
NM_201436.2 | NP_958844.1 | 3 | 289 | |||
NM_201517.2 | NP_958925.1 | 3 | 253 | |||
GenBank mRNA | AB209001.1 | - | 4 | 278 | ||
AF081192.1 | - | 4 | 232 | |||
AK023973.1 | - | 4 | 318 | |||
AK129989.1 | - | 4 | 287 | |||
AL520785.1 | - | 1 | 15 | |||
AL520811.3 | - | 4 | 268 | |||
BC000098.2 | - | 4 | 294 | 3505930 | ||
BC004274.1 | - | 3 | 219 | 3615320 | ||
BC014885.2 | - | 4 | 263 | 3907411 | ||
BC070169.1 | - | 4 | 294 | 6458566 | ||
BF242439.1 | - | 4 | 382 | |||
BG754026.1 | - | 4 | 342 | |||
BG773256.1 | - | 4 | 370 | |||
BI257891.1 | - | 4 | 325 | |||
BM452844.1 | - | 4 | 248 | |||
BM453663.1 | - | 4 | 237 | |||
DB460745.1 | - | 4 | 367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
