siRNA Details
Gene Symbol: RPS26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029.3 | NP_001020.2 | 2 | 335 | ||
GenBank mRNA | AV681946.1 | - | 2 | 335 | ||
BC002604.1 | - | 2 | 299 | 3143099 | ||
BC013215.2 | - | 1 | 45 | |||
BC015832.1 | - | 1 | 96 | 4807004 | ||
BC070220.1 | - | 1 | 96 | 6388128 | ||
BC105276.1 | - | 1 | 99 | 4287636 | ||
BC105798.1 | - | 1 | 100 | 6726218 | ||
X69654.1 | - | 1 | 93 | |||
X77770.1 | - | 1 | 70 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
