siRNA Details
Gene Symbol: AGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276343.2 | NP_001263272.2 | 8 | 1194 | ||
NM_001291379.1 | NP_001278308.1 | 10 | 1386 | |||
NM_133446.3 | NP_597703.2 | 7 | 1125 | |||
XM_011539269.2 | XP_011537571.1 | 6 | 732 | |||
XM_017015664.1 | XP_016871153.1 | 10 | 5056 | |||
XM_017015665.1 | XP_016871154.1 | 11 | 4924 | |||
XM_017015666.1 | XP_016871155.1 | 9 | 1959 | |||
XM_017015667.1 | XP_016871156.1 | 10 | 1229 | |||
XM_017015668.1 | XP_016871157.1 | 8 | 979 | |||
XM_017015669.1 | XP_016871158.1 | 8 | 887 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
