siRNA Details
Gene Symbol: AGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276343.2 | NP_001263272.2 | 8 | 2278 | ||
NM_001291379.1 | NP_001278308.1 | 10 | 2470 | |||
NM_133446.3 | NP_597703.2 | 7 | 2209 | |||
XM_011539269.2 | XP_011537571.1 | 6 | 1816 | |||
XM_017015665.1 | XP_016871154.1 | 11 | 6008 | |||
XM_017015666.1 | XP_016871155.1 | 9 | 3043 | |||
XM_017015667.1 | XP_016871156.1 | 10 | 2313 | |||
XM_017015668.1 | XP_016871157.1 | 8 | 2063 | |||
XM_017015669.1 | XP_016871158.1 | 8 | 1971 | |||
GenBank mRNA | AF411132.1 | - | 7 | 2209 | ||
BC048341.1 | - | 1 | 1331 |
Gene Symbol: AGAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133447.1 | NP_597704.1 | 12 | 2948 | ||
GenBank mRNA | AB075855.1 | - | 7 | 4352 | ||
BC140762.1 | - | 8 | 2194 | 9021630 |
Gene Symbol: AGAP7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126580.1 | 7 | 2178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
