siRNA Details
Gene Symbol: MIR4435-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024373.2 | 2 | 556 | |||
GenBank mRNA | BC014776.1 | - | 2 | 483 | ||
BC070201.1 | - | 2 | 443 |
Gene Symbol: LINC00152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024204.1 | 4 | 780 | |||
NR_024205.1 | 3 | 616 | ||||
NR_024206.1 | 2 | 470 | ||||
GenBank mRNA | BC009508.2 | - | 2 | 435 | ||
BC070202.1 | - | 393 | ||||
BM989746.1 | - | 2 | 465 | |||
CF129206.1 | - | 2 | 508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
