siRNA Details
Gene Symbol: MIR4435-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024373.2 | 2 | 528 | |||
GenBank mRNA | BC014776.1 | - | 2 | 455 | ||
BC070201.1 | - | 2 | 415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
