siRNA Details
Gene Symbol: PAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006451.4 | NP_006442.2 | 10 | 1558 | ||
NM_182789.3 | NP_877590.1 | 10 | 1321 | |||
NM_183323.2 | NP_899152.1 | 10 | 1126 | |||
XM_005248230.3 | XP_005248287.1 | 10 | 1280 | |||
XM_017008956.1 | XP_016864445.1 | 10 | 1461 | |||
GenBank mRNA | AF013758.2 | - | 10 | 1498 | ||
AK308270.1 | - | 10 | 1322 | |||
AL136920.1 | - | 10 | 1505 | |||
BC005295.1 | - | 10 | 1261 | 4052678 | ||
BC015937.2 | - | 10 | 1544 | 3922262 | ||
DQ786326.1 | - | 6 | 587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
