siRNA Details
Gene Symbol: PAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006451.4 | NP_006442.2 | 2 | 547 | ||
NM_182789.3 | NP_877590.1 | 2 | 310 | |||
NM_183323.2 | NP_899152.1 | 2 | 115 | |||
XM_005248230.3 | XP_005248287.1 | 2 | 269 | |||
XM_017008956.1 | XP_016864445.1 | 2 | 450 | |||
GenBank mRNA | AF013758.2 | - | 2 | 487 | ||
AK308270.1 | - | 2 | 310 | |||
AL136920.1 | - | 2 | 494 | |||
BC005295.1 | - | 2 | 250 | 4052678 | ||
BC015937.2 | - | 2 | 533 | 3922262 | ||
BG719757.1 | - | 2 | 87 | |||
DB089732.1 | - | 2 | 115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
