siRNA Details
Gene Symbol: MED27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253881.1 | NP_001240810.1 | 2 | 297 | ||
NM_001253882.1 | NP_001240811.1 | 2 | 297 | |||
NM_004269.3 | NP_004260.2 | 2 | 297 | |||
XM_005272236.3 | XP_005272293.1 | 2 | 276 | |||
XM_017015329.1 | XP_016870818.1 | 2 | 276 | |||
XM_017015330.1 | XP_016870819.1 | 2 | 276 | |||
GenBank mRNA | AF104252.1 | - | 2 | 140 | ||
AF230382.1 | - | 2 | 140 | |||
AK298436.1 | - | 2 | 275 | |||
AK316345.1 | - | 2 | 276 | |||
BC002878.1 | - | 2 | 284 | |||
BC064608.1 | - | 2 | 265 | 5549348 | ||
BP255072.1 | - | 2 | 297 | |||
CD388680.1 | - | 2 | 304 | |||
DQ099387.1 | - | 2 | 140 | |||
HG518166.1 | - | 2 | 275 | |||
Z36848.1 | - | 2 | 81 |
Gene Symbol: CRSP8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003665.1 | 1 | 264 | |||
GenBank mRNA | BC119722.1 | - | 2 | 271 | ||
BC119723.1 | - | 2 | 271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
