siRNA Details
Gene Symbol: RPL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000985.4 | NP_000976.1 | 7 | 905 | ||
NM_001035006.2 | NP_001030178.1 | 7 | 656 | |||
NM_001199340.1 | NP_001186269.1 | 8 | 754 | |||
NM_001199341.1 | NP_001186270.1 | 7 | 755 | |||
NM_001199342.1 | NP_001186271.1 | 8 | 853 | |||
NM_001199343.1 | NP_001186272.1 | 8 | 836 | |||
NM_001199344.1 | NP_001186273.1 | 7 | 738 | |||
NM_001199345.1 | NP_001186274.1 | 6 | 615 | |||
GenBank mRNA | AK304659.1 | - | 6 | 529 | ||
AK311828.1 | - | 7 | 570 | |||
BC000502.2 | - | 6 | 808 | 2821464 | ||
BC017831.1 | - | 1 | 574 | 4251433 | ||
BC066323.1 | - | 1 | 570 | 4797583 | ||
BC066324.1 | - | 1 | 574 | 4799029 | ||
BC106031.1 | - | 1 | 541 | 4540700 | ||
BQ882430.1 | - | 7 | 720 | |||
X53777.1 | - | 6 | 673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
