siRNA Details
Gene Symbol: YBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004559.4 | NP_004550.2 | 7 | 1118 | ||
NR_132737.1 | 6 | 936 | ||||
GenBank mRNA | AK309996.1 | - | 7 | 755 | ||
BC002411.2 | - | 7 | 901 | 2961659 | ||
BC010430.1 | - | 7 | 900 | 3506647 | ||
BC013838.1 | - | 6 | 936 | |||
BC015208.1 | - | 7 | 850 | 3914485 | ||
BC020164.1 | - | 1 | 359 | |||
BC038384.1 | - | 7 | 907 | 3449398 | ||
BC065571.1 | - | 7 | 925 | 6171221 | ||
BC070084.1 | - | 7 | 925 | 5272578 | ||
BC071708.1 | - | 7 | 910 | 4361396 | ||
BC090038.1 | - | 7 | 910 | 6526492 | ||
BC098435.1 | - | 7 | 916 | 4859074 | ||
BC106045.1 | - | 7 | 912 | 6062615 | ||
J03827.1 | - | 7 | 913 | |||
L28809.1 | - | 7 | 868 | |||
M24070.1 | - | 7 | 907 | |||
M83234.1 | - | 1 | 1009 | |||
M85234.1 | - | 1 | 1015 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
