siRNA Details
Gene Symbol: ACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097.2 | NP_001088.2 | 5 | 1187 | ||
GenBank mRNA | CR456366.1 | - | 5 | 1182 | ||
Y00970.1 | - | 5 | 1191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
