siRNA Details
Gene Symbol: RPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001256577.2 | NP_001243506.2 | 3, 4 | 445 | ||
NM_001303624.1 | NP_001290553.1 | 2, 3 | 240 | |||
NM_001303625.1 | NP_001290554.1 | 3, 4 | 449 | |||
NM_001303626.1 | NP_001290555.1 | 2, 3 | 115 | |||
NM_006013.4 | NP_006004.3 | 3, 4 | 445 | |||
GenBank mRNA | AB019572.1 | - | 1, 2 | 37 | ||
AF218023.1 | - | 1, 2 | 1004 | |||
AF486812.1 | - | 1 | 114 | |||
AK026568.1 | - | 2, 3 | 237 | |||
AK027197.1 | - | 1 | 1020 | |||
AK130582.1 | - | 3, 4 | 133 | |||
AK307511.1 | - | 3, 4 | 132 | |||
AY927570.1 | - | LIT3571 AY927570 |
1, 2 | 43 | ||
BC003358.1 | - | 2, 3 | 116 | 2900175 | ||
BC026276.1 | - | 1 | 133 | 4276627 | ||
BC071918.1 | - | 1 | 111 | 6731213 | ||
BC095425.1 | - | 1 | 123 | |||
BF667333.1 | - | 3, 4 | 280 | |||
BM761549.1 | - | 3, 4 | 137 | |||
BX647954.1 | - | 1 | 133 | |||
CA776692.1 | - | 1, 2 | 77 | |||
CR456797.1 | - | 2, 3 | 92 | |||
CR542069.1 | - | 2, 3 | 92 | |||
DQ369703.1 | - | 2, 3 | 92 | |||
DQ369704.1 | - | 2, 3 | 92 | |||
DQ369705.1 | - | 2, 3 | 92 | |||
DQ369706.1 | - | 2, 3 | 92 | |||
DQ369707.1 | - | 2, 3 | 92 | |||
DQ369708.1 | - | 2, 3 | 92 | |||
DQ369709.1 | - | 2, 3 | 92 | |||
DQ369710.1 | - | 2, 3 | 92 | |||
DQ369711.1 | - | 2, 3 | 92 | |||
DQ369712.1 | - | 2, 3 | 92 | |||
DQ369713.1 | - | 2, 3 | 92 | |||
DQ369714.1 | - | 2, 3 | 92 | |||
DQ369715.1 | - | 2, 3 | 92 | |||
DQ369716.1 | - | 2, 3 | 92 | |||
KJ534934.1 | - | 2, 3 | 92 | |||
KJ534979.1 | - | 2, 3 | 92 | |||
M64241.1 | - | 3, 4 | 133 | |||
M73791.1 | - | 2, 3 | 106 | |||
S35959.1 | - | 1, 2 | 70 | |||
S35960.1 | - | 1 | 111 |
Gene Symbol: EEF1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK292639.1 | - | 133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
