siRNA Details
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 16, 17 | 2424 | ||
NR_102404.1 | 15, 16 | 2260 | ||||
NR_102405.1 | 14, 15 | 2157 | ||||
GenBank mRNA | AY894561.1 | - | 9, 10 | 1224 | ||
AY894562.1 | - | 11, 12 | 1449 | |||
AY894563.1 | - | 10, 11 | 1323 | |||
AY894567.1 | - | 14, 15 | 2157 | |||
AY894568.1 | - | 15, 16 | 2055 | |||
AY894569.1 | - | 15, 16 | 2057 | |||
AY894570.1 | - | 11, 12 | 1449 | |||
AY894571.1 | - | 15, 16 | 2321 | |||
AY894572.1 | - | 15, 16 | 2260 | |||
AY894573.1 | - | 15, 16 | 2321 | |||
BC086308.1 | - | 16, 17 | 2267 | 5453529 |
Gene Symbol: LOC102724250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546188.2 | XP_011544490.1 | 18, 19 | 11168 |
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017940.4 | NP_060410.2 | 4071 | |||
GenBank mRNA | AB051480.1 | - | 17, 18 | 2128 | ||
AF379624.1 | - | 7, 8 | 856 | |||
AY894575.1 | - | 28, 29 | 3953 | |||
BC034418.1 | - | 12, 13 | 1633 | |||
BC094705.1 | - | 10, 11 | 1279 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 2287 | |||
NM_001302371.1 | NP_001289300.1 | 2287 | ||||
GenBank mRNA | AF379616.1 | - | 5, 6 | 834 | ||
AF379632.1 | - | 5, 6 | 545 | |||
AF379635.1 | - | 12, 13 | 1550 | |||
AF380582.1 | - | 1, 2 | 179 | |||
AL050141.1 | - | 4, 5 | 2083 | |||
AY894565.1 | - | 15, 16 | 2099 | |||
BX538005.1 | - | 24, 25 | 3534 |
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 18, 19 | 2577 | ||
NM_183372.5 | NP_899228.4 | 20, 21 | 3271 | |||
NR_046188.3 | 21, 22 | 3339 | ||||
NR_110864.1 | 19, 20 | 3263 | ||||
GenBank mRNA | AF419617.1 | - | 19, 20 | 2690 | ||
AK054850.1 | - | 4, 5 | 1016 | |||
AK302413.1 | - | 17, 18 | 2413 | |||
AL832622.2 | - | 21, 22 | 3474 | |||
BC136292.1 | - | 20, 21 | 3271 | 9020279 | ||
BX648497.1 | - | 19, 20 | 3062 |
Gene Symbol: NBPF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278141.1 | NP_001265070.1 | 27, 28 | 4282 | ||
GenBank mRNA | AB033071.1 | - | 27, 28 | 3755 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 16, 17 | 2252 | ||
XM_011509395.2 | XP_011507697.1 | 19, 20 | 2752 | |||
GenBank mRNA | AF379626.1 | - | 2, 3 | 195 | ||
AK095459.1 | - | 8, 9 | 1719 | |||
AL117237.1 | - | 10, 11 | 1249 |
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 18, 19 | 2601 | ||
NM_173638.4 | NP_775909.2 | 15, 16 | 2071 | |||
XM_017001077.1 | XP_016856566.1 | 13, 14 | 1719 | |||
GenBank mRNA | AF379630.1 | - | 10, 11 | 1249 | ||
AK097180.1 | - | 18, 19 | 2520 | |||
AK290142.1 | - | 7, 8 | 773 | |||
AK290302.1 | - | 12, 13 | 1530 | |||
AK299360.1 | - | 9, 10 | 1111 | |||
AY894566.1 | - | 12, 13 | 1511 | |||
DA932476.1 | - | 2, 3 | 205 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 13, 14 | 1712 | ||
GenBank mRNA | AF379633.1 | - | 1, 2 | 112 | ||
AF379636.1 | - | 5, 6 | 644 | |||
AK292666.1 | - | 4, 5 | 465 | |||
AK295639.1 | - | 7, 8 | 1183 |
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 16, 17 | 2282 | ||
XM_017003011.1 | XP_016858500.1 | 16, 17 | 2281 | |||
GenBank mRNA | AK294944.1 | - | 19, 20 | 2165 | ||
AW965356.1 | - | 5, 6 | 554 | |||
DA203219.1 | - | 3, 4 | 275 | |||
DA675097.1 | - | 3, 4 | 250 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277444.1 | NP_001264373.1 | 21, 22 | 2834 | ||
XM_006711317.2 | XP_006711380.1 | 22, 23 | 3062 | |||
GenBank mRNA | AK294414.1 | - | 15, 16 | 1992 | ||
DQ786323.1 | - | 4, 5 | 500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
