siRNA Details
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB051480.1 | - | 21 | 2699 | ||
BC034418.1 | - | 16 | 2204 | |||
BC094705.1 | - | 14 | 1847 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF379634.1 | - | 3 | 440 |
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 22 | 3141 | ||
NM_183372.5 | NP_899228.4 | 24 | 3835 | |||
NR_046188.3 | 25 | 3903 | ||||
NR_110864.1 | 23 | 3827 | ||||
GenBank mRNA | AK054850.1 | - | 8 | 1584 | ||
AK302413.1 | - | 21 | 2977 | |||
AL832622.2 | - | 25 | 4038 | |||
BX648497.1 | - | 23 | 3626 |
Gene Symbol: NBPF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278141.1 | NP_001265070.1 | 37 | 5582 | ||
GenBank mRNA | AF131738.1 | - | 2 | 347 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 66 | 8451 | ||
XM_011509395.2 | XP_011507697.1 | 69 | 8951 | |||
GenBank mRNA | AL117237.1 | - | 22 | 2774 |
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 22 | 3166 | ||
NM_173638.4 | NP_775909.2 | 19 | 2636 | |||
XM_017001077.1 | XP_016856566.1 | 17 | 2284 | |||
GenBank mRNA | AK290142.1 | - | 11 | 1338 | ||
AK290302.1 | - | 16 | 2095 | |||
AK299360.1 | - | 13 | 1676 | |||
AY894566.1 | - | 16 | 2076 | |||
BC023087.1 | - | 19 | 2493 | 3918932 |
Gene Symbol: NBPF25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104217.1 | 19 | 2467 | |||
GenBank mRNA | AK125792.1 | - | 19 | 2467 |
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 26 | 3582 | ||
XM_017003011.1 | XP_016858500.1 | 24 | 3356 | |||
GenBank mRNA | AK294944.1 | - | 23 | 2733 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 20 | 2992 | ||
NR_102404.1 | 19 | 2828 | ||||
NR_102405.1 | 18 | 2725 | ||||
GenBank mRNA | AK300749.1 | - | 8 | 766 | ||
AY894561.1 | - | 13 | 1795 | |||
AY894562.1 | - | 15 | 2017 | |||
AY894563.1 | - | 14 | 1891 | |||
AY894567.1 | - | 18 | 2725 | |||
AY894568.1 | - | 19 | 2623 | |||
AY894569.1 | - | 19 | 2625 | |||
AY894570.1 | - | 15 | 2017 | |||
AY894572.1 | - | 19 | 2828 | |||
AY894573.1 | - | 19 | 2889 | |||
BC086308.1 | - | 20 | 2835 | 5453529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
