siRNA Details
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 22 | 3243 | ||
NM_173638.4 | NP_775909.2 | 19 | 2713 | |||
XM_017001077.1 | XP_016856566.1 | 17 | 2361 | |||
GenBank mRNA | AK290142.1 | - | 11 | 1415 | ||
AK290302.1 | - | 16 | 2172 | |||
AK299360.1 | - | 13 | 1753 | |||
BC023087.1 | - | 19 | 2570 | 3918932 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF379634.1 | - | 3 | 517 | ||
AF419616.1 | - | 23 | 2867 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 66 | 8528 | ||
XM_011509395.2 | XP_011507697.1 | 69 | 9028 | |||
GenBank mRNA | AL117237.1 | - | 22 | 2851 |
Gene Symbol: NBPF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278267.1 | NP_001265196.1 | 16892 | |||
GenBank mRNA | AK299612.1 | - | 8 | 1066 |
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 26 | 3659 | ||
XM_017003011.1 | XP_016858500.1 | 24 | 3433 | |||
GenBank mRNA | AK294944.1 | - | 23 | 2810 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
