siRNA Details
Gene Symbol: RPL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000982.3 | NP_000973.2 | 1 | 458 | ||
GenBank mRNA | AF086172.1 | - | 1 | 357 | ||
AK130968.1 | - | 1 | 457 | |||
AK311795.1 | - | 1 | 455 | |||
BC001603.1 | - | 1 | 440 | 3543702 | ||
BC007505.2 | - | 1 | 421 | 2964441 | ||
BC019002.2 | - | 1 | 413 | |||
BC062981.1 | - | 1 | 438 | 6084268 | ||
BC070184.1 | - | 1 | 442 | 4281405 | ||
BC070323.1 | - | 1 | 455 | 4248655 | ||
BC070330.1 | - | 1 | 455 | 3931616 | ||
BC071902.1 | - | 1 | 438 | 4901492 | ||
BC104667.1 | - | 1 | 459 | 6724053 | ||
BX647669.1 | - | 1 | 457 | |||
CR457032.1 | - | 1 | 415 | |||
U14967.1 | - | 1 | 444 | |||
U25789.1 | - | 1 | 448 | |||
X89401.1 | - | 1 | 428 |
Gene Symbol: RPL21P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209100.1 | - | 1 | 517 |
Gene Symbol: RPL21P28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026911.1 | 1 | 460 |

More Information
Other Species Targeted

Gene Ontology Categories:
