siRNA Details
Gene Symbol: PRELID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271828.1 | NP_001258757.1 | 1 | 304 | ||
NM_013237.3 | NP_037369.1 | 1 | 304 | |||
GenBank mRNA | AF087858.1 | - | 1 | 250 | ||
AF111112.1 | - | 1 | 253 | |||
AF112203.1 | - | 1 | 248 | |||
AF151864.1 | - | 1 | 256 | |||
AF153607.1 | - | 1 | 238 | |||
AF201925.1 | - | 1 | 122 | |||
AK312170.1 | - | 1 | 258 | |||
BC000007.2 | - | 1 | 253 | 3505068 | ||
BC007268.1 | - | 1 | 267 | 3139416 | ||
BC008307.1 | - | 1 | 230 | 3606229 | ||
BC008866.2 | - | 1 | 253 | 4128831 | ||
BC013733.1 | - | 1 | 245 | 3862833 | ||
BC013748.1 | - | 1 | 232 | 3867564 | ||
BC018904.2 | - | 1 | 267 | |||
BG739785.1 | - | 1 | 304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
