siRNA Details
Gene Symbol: PRELID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013237.3 | NP_037369.1 | 5 | 759 | ||
GenBank mRNA | AF087858.1 | - | 5 | 705 | ||
AF111112.1 | - | 1 | 708 | |||
AF151864.1 | - | 5 | 711 | |||
AF153607.1 | - | 5 | 693 | |||
AF201925.1 | - | 5 | 577 | |||
AK312170.1 | - | 5 | 713 | |||
BC000007.2 | - | 5 | 708 | 3505068 | ||
BC007268.1 | - | 5 | 722 | 3139416 | ||
BC008307.1 | - | 5 | 685 | 3606229 | ||
BC008866.2 | - | 5 | 708 | 4128831 | ||
BC013733.1 | - | 5 | 700 | 3862833 | ||
BC013748.1 | - | 5 | 687 | 3867564 | ||
BC018904.2 | - | 5 | 722 | |||
BG739785.1 | - | 5 | 755 | |||
BU956915.1 | - | 4 | 310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
