siRNA Details
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006908.4 | NP_008839.2 | 4 | 488 | ||
NM_018890.3 | NP_061485.1 | 5 | 545 | |||
GenBank mRNA | AB451238.1 | - | 4 | 247 | ||
AB451363.1 | - | 4 | 247 | |||
AF136373.1 | - | 5 | 304 | |||
AF498964.1 | - | 4 | 247 | |||
AJ132694.1 | - | 5 | 304 | |||
AK054993.1 | - | 4 | 463 | |||
AK291806.1 | - | 4 | 444 | |||
AK291815.1 | - | 5 | 491 | |||
AY279384.1 | - | 4 | 405 | |||
BC004247.1 | - | 4 | 385 | 3610745 | ||
BC050687.1 | - | 4 | 435 | 6149377 | ||
BC107748.1 | - | 4 | 450 | 6699945 | ||
BQ689587.1 | - | 5 | 504 | |||
BT007121.1 | - | 4 | 247 | |||
M29870.1 | - | 4 | 247 | |||
M31467.1 | - | 4 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
