siRNA Details
Gene Symbol: IMPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144878.1 | NP_001138350.1 | 3 | 345 | ||
NM_001144879.1 | NP_001138351.1 | 2 | 144 | |||
NM_005536.3 | NP_005527.1 | 2 | 144 | |||
GenBank mRNA | AF042729.2 | - | 2 | 115 | ||
AK297078.1 | - | 2 | 91 | |||
AK300750.1 | - | 3 | 274 | |||
AK312823.1 | - | 2 | 90 | |||
BC008381.1 | - | 2 | 110 | 3682305 | ||
BC009565.1 | - | 2 | 114 | 3682657 | ||
BP270804.1 | - | 3 | 324 | |||
DC375686.1 | - | 2 | 145 | |||
X66922.1 | - | 1 | 53 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
