siRNA Details
Gene Symbol: POU5F1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159542.1 | NP_001153014.1 | 1 | 890 | ||
GenBank mRNA | AF268615.1 | - | 1 | 635 | ||
DQ486513.1 | - | 1 | 890 | |||
GU480835.1 | - | 1 | 636 | |||
GU480840.1 | - | 1 | 636 | |||
GU480841.1 | - | 1 | 636 | |||
GU480842.1 | - | 1 | 636 | |||
GU480844.1 | - | 1 | 636 | |||
GU480845.1 | - | 1 | 636 | |||
GU480849.1 | - | 1 | 638 | |||
GU480850.1 | - | 1 | 637 | |||
GU480851.1 | - | 1 | 637 | |||
GU480864.1 | - | 1 | 636 | |||
GU480868.1 | - | 1 | 636 | |||
GU480884.1 | - | 1 | 635 | |||
GU480885.1 | - | 1 | 637 | |||
GU480886.1 | - | 1 | 636 | |||
GU480896.1 | - | 1 | 636 | |||
GU480899.1 | - | 1 | 637 | |||
GU480902.1 | - | 1 | 636 | |||
GU480905.1 | - | 1 | 636 | |||
GU480906.1 | - | 1 | 636 | |||
GU480908.1 | - | 1 | 636 | |||
GU480910.1 | - | 1 | 638 | |||
GU480911.1 | - | 1 | 636 | |||
GU480912.1 | - | 1 | 636 | |||
GU480913.1 | - | 1 | 636 | |||
HQ122710.1 | - | 1 | 636 | |||
HQ122711.1 | - | 1 | 636 | |||
HQ122712.1 | - | 1 | 636 | |||
HQ122715.1 | - | 1 | 635 | |||
HQ122717.1 | - | 1 | 636 | |||
HQ122721.1 | - | 1 | 636 | |||
HQ122749.1 | - | 1 | 636 | |||
HQ122781.1 | - | 1 | 636 | |||
HQ122782.1 | - | 1 | 636 | |||
HQ122783.1 | - | 1 | 636 | |||
HQ122786.1 | - | 1 | 636 | |||
HQ122787.1 | - | 1 | 636 | |||
HQ122816.1 | - | 1 | 636 | |||
HQ122817.1 | - | 1 | 636 | |||
HQ122818.1 | - | 1 | 636 | |||
HQ122819.1 | - | 1 | 636 | |||
HQ122820.1 | - | 1 | 636 | |||
HQ122821.1 | - | 1 | 636 | |||
HQ122822.1 | - | 1 | 636 | |||
HQ122823.1 | - | 1 | 636 | |||
HQ122824.1 | - | 1 | 636 | |||
HQ122825.1 | - | 1 | 636 | |||
HQ122826.1 | - | 1 | 636 | |||
HQ122827.1 | - | 1 | 636 | |||
HQ122841.1 | - | 1 | 636 | |||
HQ122842.1 | - | 1 | 636 | |||
HQ122843.1 | - | 1 | 636 | |||
HQ122844.1 | - | 1 | 636 | |||
HQ122845.1 | - | 1 | 636 | |||
HQ122846.1 | - | 1 | 636 | |||
HQ122847.1 | - | 1 | 636 | |||
HQ122848.1 | - | 1 | 636 | |||
HQ122849.1 | - | 1 | 636 | |||
HQ122850.1 | - | 1 | 637 | |||
HQ122851.1 | - | 1 | 636 | |||
HQ122852.1 | - | 1 | 636 | |||
HQ122853.1 | - | 1 | 636 | |||
HQ122854.1 | - | 1 | 636 | |||
HQ122855.1 | - | 1 | 636 | |||
HQ122856.1 | - | 1 | 636 | |||
HQ122857.1 | - | 1 | 636 | |||
HQ122863.1 | - | 1 | 636 | |||
HQ122864.1 | - | 1 | 636 | |||
HQ122865.1 | - | 1 | 636 | |||
HQ122866.1 | - | 1 | 636 | |||
HQ122867.1 | - | 1 | 636 | |||
HQ122868.1 | - | 1 | 636 | |||
HQ122869.1 | - | 1 | 636 | |||
HQ122870.1 | - | 1 | 636 | |||
HQ122871.1 | - | 1 | 636 | |||
HQ122873.1 | - | 1 | 635 | |||
HQ122874.1 | - | 1 | 636 | |||
HQ122875.1 | - | 1 | 636 | |||
HQ122876.1 | - | 1 | 636 | |||
HQ122877.1 | - | 1 | 636 | |||
HQ122878.1 | - | 1 | 636 | |||
HQ122879.1 | - | 1 | 636 | |||
HQ122880.1 | - | 1 | 636 | |||
HQ122881.1 | - | 1 | 636 | |||
HQ122882.1 | - | 1 | 636 | |||
HQ122883.1 | - | 1 | 636 | |||
HQ122884.1 | - | 1 | 636 | |||
HQ122885.1 | - | 1 | 636 | |||
HQ122886.1 | - | 1 | 636 | |||
HQ122887.1 | - | 1 | 636 | |||
HQ122893.1 | - | 1 | 636 | |||
HQ122903.1 | - | 1 | 636 |
Gene Symbol: POU5F1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036440.1 | 1 | 634 | |||
GenBank mRNA | AF268617.1 | - | 1 | 633 | ||
GU480832.1 | - | 1 | 634 | |||
GU480837.1 | - | 1 | 634 | |||
GU480838.1 | - | 1 | 634 | |||
GU480843.1 | - | 1 | 634 | |||
GU480846.1 | - | 1 | 634 | |||
GU480876.1 | - | 1 | 634 | |||
GU480877.1 | - | 1 | 634 | |||
GU480887.1 | - | 1 | 634 | |||
GU480888.1 | - | 1 | 635 | |||
GU480891.1 | - | 1 | 635 | |||
GU480892.1 | - | 1 | 634 | |||
GU480893.1 | - | 1 | 634 | |||
GU480897.1 | - | 1 | 634 | |||
GU480901.1 | - | 1 | 633 | |||
GU480903.1 | - | 1 | 634 | |||
GU480907.1 | - | 1 | 634 | |||
GU480909.1 | - | 1 | 634 | |||
HQ122692.1 | - | 1 | 634 | |||
HQ122693.1 | - | 1 | 634 | |||
HQ122694.1 | - | 1 | 634 | |||
HQ122722.1 | - | 1 | 634 | |||
HQ122723.1 | - | 1 | 634 | |||
HQ122724.1 | - | 1 | 634 | |||
HQ122725.1 | - | 1 | 634 | |||
HQ122726.1 | - | 1 | 634 | |||
HQ122727.1 | - | 1 | 634 | |||
HQ122728.1 | - | 1 | 634 | |||
HQ122729.1 | - | 1 | 634 | |||
HQ122730.1 | - | 1 | 634 | |||
HQ122753.1 | - | 1 | 634 | |||
HQ122754.1 | - | 1 | 634 | |||
HQ122755.1 | - | 1 | 634 | |||
HQ122791.1 | - | 1 | 634 | |||
HQ122792.1 | - | 1 | 634 | |||
HQ122793.1 | - | 1 | 634 | |||
HQ122794.1 | - | 1 | 634 | |||
HQ122795.1 | - | 1 | 634 | |||
HQ122796.1 | - | 1 | 634 | |||
HQ122797.1 | - | 1 | 634 | |||
HQ122828.1 | - | 1 | 634 | |||
HQ122829.1 | - | 1 | 634 | |||
HQ122830.1 | - | 1 | 634 | |||
HQ122831.1 | - | 1 | 634 | |||
Z21964.1 | - | 1 | 75 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
