siRNA Details
Gene Symbol: POU5F1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036440.1 | 1 | 734 | |||
GenBank mRNA | AF268617.1 | - | 1 | 733 | ||
GU480832.1 | - | 1 | 734 | |||
GU480837.1 | - | 1 | 734 | |||
GU480838.1 | - | 1 | 734 | |||
GU480843.1 | - | 1 | 734 | |||
GU480846.1 | - | 1 | 734 | |||
GU480847.1 | - | 1 | 734 | |||
GU480876.1 | - | 1 | 734 | |||
GU480877.1 | - | 1 | 734 | |||
GU480887.1 | - | 1 | 734 | |||
GU480888.1 | - | 1 | 735 | |||
GU480891.1 | - | 1 | 735 | |||
GU480892.1 | - | 1 | 734 | |||
GU480893.1 | - | 1 | 734 | |||
GU480897.1 | - | 1 | 734 | |||
GU480903.1 | - | 1 | 734 | |||
GU480907.1 | - | 1 | 734 | |||
GU480909.1 | - | 1 | 734 | |||
GU480915.1 | - | 1 | 734 | |||
HQ122722.1 | - | 1 | 734 | |||
HQ122723.1 | - | 1 | 734 | |||
HQ122724.1 | - | 1 | 734 | |||
HQ122725.1 | - | 1 | 734 | |||
HQ122726.1 | - | 1 | 734 | |||
HQ122727.1 | - | 1 | 734 | |||
HQ122728.1 | - | 1 | 734 | |||
HQ122729.1 | - | 1 | 734 | |||
HQ122730.1 | - | 1 | 734 | |||
HQ122753.1 | - | 1 | 734 | |||
HQ122754.1 | - | 1 | 734 | |||
HQ122755.1 | - | 1 | 734 | |||
HQ122791.1 | - | 1 | 734 | |||
HQ122792.1 | - | 1 | 734 | |||
HQ122793.1 | - | 1 | 734 | |||
HQ122794.1 | - | 1 | 734 | |||
HQ122795.1 | - | 1 | 734 | |||
HQ122796.1 | - | 1 | 734 | |||
HQ122797.1 | - | 1 | 734 | |||
HQ122828.1 | - | 1 | 734 | |||
HQ122829.1 | - | 1 | 734 | |||
HQ122830.1 | - | 1 | 734 | |||
HQ122831.1 | - | 1 | 734 | |||
Z21964.1 | - | 1 | 175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
