siRNA Details
Gene Symbol: METTL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181725.3 | NP_859076.3 | 2 | 181 | ||
XM_017024572.1 | XP_016880061.1 | 2 | 169 | |||
GenBank mRNA | AF161384.1 | - | 2 | 159 | ||
AF161385.1 | - | 129 | ||||
AK022822.1 | - | 2 | 158 | |||
BC006985.1 | - | 1 | 383 | 3930328 | ||
BF975211.1 | - | 2 | 154 | |||
BP255803.1 | - | 2 | 181 | |||
DA027807.1 | - | 2 | 169 |
Gene Symbol: METTL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018396.2 | NP_060866.2 | 2 | 200 | ||
GenBank mRNA | AK299960.1 | - | 2 | 164 | ||
AK302775.1 | - | 2 | 181 | |||
BC064929.1 | - | 2 | 160 | |||
BC121115.1 | - | 2 | 125 | |||
BC121116.1 | - | 2 | 125 | |||
BP247182.1 | - | 2 | 200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
