siRNA Details
Gene Symbol: METTL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018396.2 | NP_060866.2 | 3 | 276 | ||
GenBank mRNA | AK002212.1 | - | 2 | 149 | ||
AK299960.1 | - | 3 | 240 | |||
AK302775.1 | - | 3 | 257 | |||
BC064929.1 | - | 3 | 236 | |||
BC107586.1 | - | 2 | 160 | 6494734 | ||
BC121115.1 | - | 3 | 201 | |||
BC121116.1 | - | 3 | 201 | |||
BP247182.1 | - | 3 | 276 |
Gene Symbol: METTL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181725.3 | NP_859076.3 | 3 | 257 | ||
XM_017024572.1 | XP_016880061.1 | 3 | 245 | |||
GenBank mRNA | AF161384.1 | - | 3 | 235 | ||
AF161385.1 | - | 205 | ||||
AK022822.1 | - | 3 | 234 | |||
BC006985.1 | - | 2 | 459 | 3930328 | ||
BF975211.1 | - | 3 | 230 | |||
BP255803.1 | - | 3 | 258 | |||
DA027807.1 | - | 3 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
