siRNA Details
Gene Symbol: FAM21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005751.2 | NP_001005751.1 | 29 | 3759 | ||
NM_001291398.1 | NP_001278327.1 | 28 | 3696 | |||
XM_005269806.3 | XP_005269863.2 | 28 | 3635 | |||
XM_005269807.3 | XP_005269864.2 | 28 | 3620 | |||
XM_005269808.3 | XP_005269865.2 | 27 | 3521 | |||
XM_005269810.3 | XP_005269867.2 | 11 | 1852 | |||
XM_006717831.3 | XP_006717894.2 | 28 | 3554 | |||
XM_006717832.3 | XP_006717895.2 | 26 | 3419 | |||
XM_011539786.2 | XP_011538088.1 | 26 | 3308 | |||
XM_017016227.1 | XP_016871716.1 | 28 | 3584 | |||
XM_017016228.1 | XP_016871717.1 | 27 | 3572 | |||
XM_017016229.1 | XP_016871718.1 | 27 | 3557 | |||
XM_017016230.1 | XP_016871719.1 | 28 | 3572 | |||
XM_017016231.1 | XP_016871720.1 | 27 | 3491 | |||
XM_017016232.1 | XP_016871721.1 | 26 | 3485 | |||
XM_017016233.1 | XP_016871722.1 | 26 | 3282 | |||
XM_017016234.1 | XP_016871723.1 | 28 | 3642 | |||
XM_017016235.1 | XP_016871724.1 | 20 | 2658 | |||
GenBank mRNA | AK125147.1 | - | 1 | 2113 | ||
AK304116.1 | - | 29 | 3695 | |||
AL080183.1 | - | 5 | 1091 | |||
BC075815.1 | - | 25 | 3261 | |||
BC082258.1 | - | 29 | 3686 | 6278433 | ||
BC144479.1 | - | 28 | 3725 | 9053006 | ||
BQ060891.1 | - | 1 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
