siRNA Details
Gene Symbol: POU5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002701.5 | NP_002692.2 | 1 | 91 | ||
GenBank mRNA | BC117435.1 | - | 1 | 72 | 40125986 | |
CD247724.1 | - | 1 | 114 | |||
GU480826.1 | - | 1 | 19 | |||
GU480829.1 | - | 1 | 19 | |||
GU480830.1 | - | 1 | 19 | |||
HQ122658.1 | - | 1 | 19 | |||
HQ122659.1 | - | 1 | 19 | |||
HQ122660.1 | - | 1 | 19 | |||
HQ122661.1 | - | 1 | 19 | |||
HQ122662.1 | - | 1 | 19 | |||
HQ122663.1 | - | 1 | 19 | |||
HQ122664.1 | - | 1 | 19 | |||
HQ122665.1 | - | 1 | 19 | |||
HQ122666.1 | - | 1 | 19 | |||
HQ122667.1 | - | 1 | 19 | |||
HQ122668.1 | - | 1 | 19 | |||
HQ122669.1 | - | 1 | 19 | |||
HQ122670.1 | - | 1 | 19 | |||
HQ122671.1 | - | 1 | 19 | |||
HQ122672.1 | - | 1 | 19 | |||
HQ122673.1 | - | 1 | 19 | |||
HQ122674.1 | - | 1 | 19 | |||
HQ122675.1 | - | 1 | 19 | |||
Z11898.1 | - | 1 | 60 |
Gene Symbol: POU5F1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036440.1 | 1 | 19 | |||
GenBank mRNA | AF268617.1 | - | 1 | 18 | ||
GU480832.1 | - | 1 | 19 | |||
GU480837.1 | - | 1 | 19 | |||
GU480838.1 | - | 1 | 19 | |||
GU480843.1 | - | 1 | 19 | |||
GU480846.1 | - | 1 | 19 | |||
GU480847.1 | - | 1 | 19 | |||
GU480876.1 | - | 1 | 19 | |||
GU480877.1 | - | 1 | 19 | |||
GU480887.1 | - | 1 | 19 | |||
GU480891.1 | - | 1 | 19 | |||
GU480892.1 | - | 1 | 19 | |||
GU480893.1 | - | 1 | 19 | |||
GU480897.1 | - | 1 | 19 | |||
GU480901.1 | - | 1 | 19 | |||
GU480903.1 | - | 1 | 19 | |||
GU480907.1 | - | 1 | 19 | |||
GU480909.1 | - | 1 | 19 | |||
GU480915.1 | - | 1 | 19 | |||
HQ122692.1 | - | 1 | 19 | |||
HQ122693.1 | - | 1 | 19 | |||
HQ122694.1 | - | 1 | 19 | |||
HQ122722.1 | - | 1 | 19 | |||
HQ122723.1 | - | 1 | 19 | |||
HQ122724.1 | - | 1 | 19 | |||
HQ122725.1 | - | 1 | 19 | |||
HQ122726.1 | - | 1 | 19 | |||
HQ122727.1 | - | 1 | 19 | |||
HQ122728.1 | - | 1 | 19 | |||
HQ122729.1 | - | 1 | 19 | |||
HQ122730.1 | - | 1 | 19 | |||
HQ122753.1 | - | 1 | 19 | |||
HQ122754.1 | - | 1 | 19 | |||
HQ122755.1 | - | 1 | 19 | |||
HQ122791.1 | - | 1 | 19 | |||
HQ122792.1 | - | 1 | 19 | |||
HQ122793.1 | - | 1 | 19 | |||
HQ122794.1 | - | 1 | 19 | |||
HQ122795.1 | - | 1 | 19 | |||
HQ122796.1 | - | 1 | 19 | |||
HQ122797.1 | - | 1 | 19 | |||
HQ122828.1 | - | 1 | 19 | |||
HQ122829.1 | - | 1 | 19 | |||
HQ122830.1 | - | 1 | 19 | |||
HQ122831.1 | - | 1 | 19 |
Gene Symbol: POU5F1P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GU480890.1 | - | 1 | 19 | ||
GU480900.1 | - | 1 | 19 | |||
GU480914.1 | - | 1 | 19 | |||
HQ122695.1 | - | 1 | 19 | |||
HQ122696.1 | - | 1 | 19 | |||
HQ122697.1 | - | 1 | 19 | |||
HQ122707.1 | - | 1 | 19 | |||
HQ122731.1 | - | 1 | 19 | |||
HQ122745.1 | - | 1 | 19 | |||
HQ122747.1 | - | 1 | 19 | |||
HQ122906.1 | - | 1 | 19 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
