siRNA Details
Gene Symbol: POU5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002701.5 | NP_002692.2 | 1 | 174 | ||
GenBank mRNA | AM851115.1 | - | 1 | 147 | ||
AM851116.1 | - | 1 | 147 | |||
AM851117.1 | - | 1 | 148 | |||
AM851118.1 | - | 1 | 150 | |||
AM851119.1 | - | 1 | 148 | |||
AM851120.1 | - | 1 | 148 | |||
BC117435.1 | - | 1 | 155 | 40125986 | ||
BC117437.1 | - | 1 | 155 | 40125988 | ||
BQ278003.1 | - | 157 | ||||
CD247724.1 | - | 1 | 197 | |||
GU480808.1 | - | 1 | 102 | |||
GU480809.1 | - | 1 | 102 | |||
GU480810.1 | - | 1 | 102 | |||
GU480811.1 | - | 1 | 102 | |||
GU480812.1 | - | 1 | 102 | |||
GU480813.1 | - | 1 | 102 | |||
GU480815.1 | - | 1 | 102 | |||
GU480817.1 | - | 1 | 102 | |||
GU480818.1 | - | 1 | 102 | |||
GU480819.1 | - | 1 | 102 | |||
GU480820.1 | - | 1 | 102 | |||
GU480821.1 | - | 1 | 102 | |||
GU480822.1 | - | 1 | 102 | |||
GU480823.1 | - | 1 | 102 | |||
GU480824.1 | - | 1 | 102 | |||
GU480825.1 | - | 1 | 102 | |||
GU480826.1 | - | 1 | 102 | |||
GU480827.1 | - | 1 | 102 | |||
GU480828.1 | - | 1 | 102 | |||
GU480829.1 | - | 1 | 102 | |||
GU480830.1 | - | 1 | 102 | |||
HQ122658.1 | - | 1 | 102 | |||
HQ122659.1 | - | 1 | 102 | |||
HQ122660.1 | - | 1 | 102 | |||
HQ122661.1 | - | 1 | 102 | |||
HQ122662.1 | - | 1 | 102 | |||
HQ122663.1 | - | 1 | 102 | |||
HQ122664.1 | - | 1 | 102 | |||
HQ122665.1 | - | 1 | 102 | |||
HQ122666.1 | - | 1 | 102 | |||
HQ122667.1 | - | 1 | 102 | |||
HQ122668.1 | - | 1 | 102 | |||
HQ122669.1 | - | 1 | 102 | |||
HQ122670.1 | - | 1 | 102 | |||
HQ122671.1 | - | 1 | 102 | |||
HQ122672.1 | - | 1 | 102 | |||
HQ122673.1 | - | 1 | 102 | |||
HQ122674.1 | - | 1 | 102 | |||
HQ122675.1 | - | 1 | 102 | |||
Z11898.1 | - | 1 | 143 |
Gene Symbol: POU5F1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GU480886.1 | - | 1 | 102 |
Gene Symbol: POU5F1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036440.1 | 1 | 102 | |||
GenBank mRNA | AF268617.1 | - | 1 | 101 | ||
GU480837.1 | - | 1 | 102 | |||
GU480838.1 | - | 1 | 102 | |||
GU480843.1 | - | 1 | 102 | |||
GU480846.1 | - | 1 | 102 | |||
GU480847.1 | - | 1 | 102 | |||
GU480876.1 | - | 1 | 102 | |||
GU480877.1 | - | 1 | 102 | |||
GU480887.1 | - | 1 | 102 | |||
GU480888.1 | - | 1 | 103 | |||
GU480891.1 | - | 1 | 102 | |||
GU480892.1 | - | 1 | 102 | |||
GU480893.1 | - | 1 | 102 | |||
GU480897.1 | - | 1 | 102 | |||
GU480901.1 | - | 1 | 102 | |||
GU480903.1 | - | 1 | 102 | |||
GU480907.1 | - | 1 | 102 | |||
GU480909.1 | - | 1 | 102 | |||
HQ122692.1 | - | 1 | 102 | |||
HQ122693.1 | - | 1 | 102 | |||
HQ122694.1 | - | 1 | 102 | |||
HQ122722.1 | - | 1 | 102 | |||
HQ122723.1 | - | 1 | 102 | |||
HQ122724.1 | - | 1 | 102 | |||
HQ122725.1 | - | 1 | 102 | |||
HQ122726.1 | - | 1 | 102 | |||
HQ122727.1 | - | 1 | 102 | |||
HQ122728.1 | - | 1 | 102 | |||
HQ122729.1 | - | 1 | 102 | |||
HQ122730.1 | - | 1 | 102 | |||
HQ122753.1 | - | 1 | 102 | |||
HQ122754.1 | - | 1 | 102 | |||
HQ122755.1 | - | 1 | 102 | |||
HQ122791.1 | - | 1 | 102 | |||
HQ122792.1 | - | 1 | 102 | |||
HQ122793.1 | - | 1 | 102 | |||
HQ122794.1 | - | 1 | 102 | |||
HQ122795.1 | - | 1 | 102 | |||
HQ122796.1 | - | 1 | 102 | |||
HQ122797.1 | - | 1 | 102 | |||
HQ122828.1 | - | 1 | 102 | |||
HQ122829.1 | - | 1 | 102 | |||
HQ122830.1 | - | 1 | 102 | |||
HQ122831.1 | - | 1 | 102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
