siRNA Details
Gene Symbol: FTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002032.2 | NP_002023.2 | 2 | 441 | ||
GenBank mRNA | AB062402.1 | - | 2 | 394 | ||
AF088851.1 | - | 2 | 248 | |||
AK311818.1 | - | 2 | 412 | |||
AY258285.1 | - | 999 | ||||
BC000857.1 | - | 2 | 400 | 3459353 | ||
BC001399.2 | - | 2 | 369 | 3051458 | ||
BC011359.2 | - | 2 | 391 | 4157296 | ||
BC013724.2 | - | 2 | 394 | 3857790 | ||
BC015156.1 | - | 2 | 264 | 3883694 | ||
BC016009.1 | - | 2 | 416 | 4666545 | ||
BC016857.1 | - | 2 | 360 | 3848510 | ||
BC020300.1 | - | 2 | 377 | |||
BC035441.1 | - | 2 | 400 | |||
BC063514.1 | - | 2 | 383 | 3882345 | ||
BC066341.1 | - | 3 | 318 | 5288688 | ||
BC066961.1 | - | 2 | 414 | 4796312 | ||
BC070494.1 | - | 3 | 355 | |||
BC073750.1 | - | 2 | 398 | 6256153 | ||
BC104643.1 | - | 2 | 416 | 4043391 | ||
BC105802.1 | - | 3 | 393 | |||
BQ887222.1 | - | 441 | ||||
L20941.1 | - | 2 | 414 | |||
M11146.1 | - | 2 | 283 | |||
M12937.1 | - | 2 | 232 | |||
M97164.1 | - | 2 | 335 | |||
X00318.1 | - | 2 | 297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
