siRNA Details
Gene Symbol: FBXW10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267585.1 | NP_001254514.1 | 14 | 2852 | ||
NM_001267586.1 | NP_001254515.1 | 13 | 2693 | |||
GenBank mRNA | AL136822.1 | - | 13 | 2631 | ||
AY729024.1 | - | 14 | 2620 | |||
BC028364.2 | - | 12 | 2866 | 4827344 |
Gene Symbol: CDRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282540.1 | NP_001269469.1 | 13 | 2620 | ||
GenBank mRNA | AF337810.1 | - | 1 | 205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
