siRNA Details
Gene Symbol: FAM3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040020.1 | NP_001035109.1 | 7 | 568 | ||
NM_014888.2 | NP_055703.1 | 7 | 608 | |||
XM_011515736.2 | XP_011514038.1 | 8 | 669 | |||
XM_011515737.2 | XP_011514039.1 | 8 | 700 | |||
GenBank mRNA | AK290682.1 | - | 7 | 563 | ||
AK292731.1 | - | 7 | 548 | |||
BC024200.1 | - | 7 | 541 | 3532209 | ||
BC046932.1 | - | 7 | 536 | 4396117 | ||
BC068526.1 | - | 7 | 574 | 5287680 | ||
D87120.1 | - | 1 | 527 | |||
DA144459.1 | - | 7 | 507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
