siRNA Details
Gene Symbol: FAM3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040020.1 | NP_001035109.1 | 9, 10 | 807 | ||
NM_014888.2 | NP_055703.1 | 9, 10 | 847 | |||
XM_011515736.2 | XP_011514038.1 | 10, 11 | 908 | |||
XM_011515737.2 | XP_011514039.1 | 10, 11 | 939 | |||
GenBank mRNA | AK290682.1 | - | 9, 10 | 802 | ||
AK292731.1 | - | 9, 10 | 787 | |||
BC024200.1 | - | 9, 10 | 780 | 3532209 | ||
BC046932.1 | - | 9, 10 | 775 | 4396117 | ||
BC068526.1 | - | 9, 10 | 813 | 5287680 | ||
D87120.1 | - | 2 | 766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
