siRNA Details
Gene Symbol: AHCTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323342.1 | NP_001310271.1 | 3 | 644 | ||
NM_001323343.1 | NP_001310272.1 | 3 | 644 | |||
NM_015446.4 | NP_056261.4 | 3 | 360 | |||
NR_136586.1 | 3 | 644 | ||||
XM_006711758.1 | XP_006711821.1 | 3 | 473 | |||
XM_011544156.2 | XP_011542458.2 | 3 | 339 | |||
XM_011544157.2 | XP_011542459.2 | 3 | 339 | |||
GenBank mRNA | AB059277.1 | - | 3 | 360 | ||
AY157619.1 | - | 3 | 478 |
Gene Symbol: AHCTF1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077058.1 | 1 | 460 | |||
GenBank mRNA | AK057394.1 | - | 1 | 294 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
