siRNA Details
Gene Symbol: AHCTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323342.1 | NP_001310271.1 | 4 | 795 | ||
NM_001323343.1 | NP_001310272.1 | 4 | 795 | |||
NM_015446.4 | NP_056261.4 | 4 | 511 | |||
NR_136586.1 | 4 | 795 | ||||
XM_006711758.1 | XP_006711821.1 | 4 | 624 | |||
XM_011544156.2 | XP_011542458.2 | 4 | 490 | |||
XM_011544157.2 | XP_011542459.2 | 4 | 490 | |||
GenBank mRNA | AB059277.1 | - | 4 | 511 | ||
AY157619.1 | - | 4 | 629 |
Gene Symbol: AHCTF1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077058.1 | 1 | 611 | |||
GenBank mRNA | AK057394.1 | - | 1 | 445 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
