siRNA Details
Gene Symbol: RPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253379.1 | NP_001240308.1 | 4 | 770 | ||
NM_001253380.1 | NP_001240309.1 | 3 | 532 | |||
NM_001253382.1 | NP_001240311.1 | 4 | 521 | |||
NM_001253383.1 | NP_001240312.1 | 4 | 500 | |||
NM_002948.3 | NP_002939.2 | 4 | 757 | |||
GenBank mRNA | AF279903.1 | - | 3 | 411 | ||
AF283772.2 | - | 4 | 417 | |||
AK091751.1 | - | 2 | 1497 | |||
AK098795.1 | - | 4 | 407 | |||
AK129815.1 | - | 4 | 417 | |||
AK290694.1 | - | 4 | 413 | |||
AK292000.1 | - | 4 | 413 | |||
AY347528.1 | - | 3 | 482 | |||
BC014837.1 | - | 4 | 420 | 2901238 | ||
BC068198.1 | - | 1 | 416 | 4093343 | ||
BC070328.1 | - | 4 | 413 | 30371842 | ||
BC071672.1 | - | 1 | 402 | 4390593 | ||
BE893727.1 | - | 4 | 492 | |||
BT019672.1 | - | 3 | 377 | |||
BU178524.1 | - | 4 | 521 | |||
BX537495.1 | - | 2 | 1440 | |||
DA037869.1 | - | 4 | 438 | |||
DA205054.1 | - | 4 | 483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
