siRNA Details
Gene Symbol: RPL27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000990.4 | NP_000981.1 | 4 | 588 | ||
GenBank mRNA | AK311767.1 | - | 4 | 244 | ||
BC005326.1 | - | 1 | 244 | 4052417 | ||
BC020169.1 | - | 3 | 220 | |||
BC066326.1 | - | 1 | 244 | 5311530 | ||
BG687024.1 | - | 278 | ||||
BQ058236.1 | - | 4 | 266 | |||
U14968.1 | - | 4 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
