siRNA Details
Gene Symbol: RPS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011.3 | NP_001002.1 | 6 | 577 | ||
GenBank mRNA | AB209386.1 | - | 2 | 4735 | ||
AK092027.1 | - | 1 | 2434 | |||
AK311794.1 | - | 6 | 515 | |||
BC002866.2 | - | 6 | 533 | 3940495 | ||
BC061901.1 | - | 6 | 570 | 4442840 | ||
BC071919.1 | - | 1 | 497 | 5811776 | ||
M77233.1 | - | 5 | 466 | |||
X74803.1 | - | 1 | 84 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
