siRNA Details
Gene Symbol: SPCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014752.2 | NP_055567.2 | 4 | 451 | ||
GenBank mRNA | AK293128.1 | - | 4 | 432 | ||
BC008063.2 | - | 1 | 312 | 2967488 | ||
BC064957.1 | - | 4 | 416 | 6092500 | ||
BC070276.1 | - | 1 | 189 | 4656138 | ||
BC082231.1 | - | 1 | 306 | 3954096 | ||
BC106066.1 | - | 4 | 416 | 4521501 | ||
BP338843.1 | - | 4 | 451 | |||
CR542233.1 | - | 2 | 103 | |||
CR542243.1 | - | 2 | 103 | |||
D14658.1 | - | 4 | 410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
