siRNA Details
Gene Symbol: GJC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181538.2 | NP_853516.1 | 1 | 502 | ||
GenBank mRNA | AF503615.1 | - | 1 | 502 | ||
AY297109.1 | - | 1 | 502 | |||
BC043381.1 | - | 1 | 146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
